α-Benzoinoxime 97%

Supplier: Ambeed
Danger

A286757-10G A286757-5G
76693-150EA 9 USD
76693-150 76693-152
α-Benzoinoxime 97%
α-Benzoinoxime

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₂
MDL Number: MFCD00004501
CAS Number: 441-38-3
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR