Thionin(acetate) 85% (dye content)

Supplier: Ambeed
Warning

A286246-1G A286246-250MG A286246-5G
76693-132EA 25 USD
76693-132 76693-134 76693-136
Thionin(acetate) 85% (dye content)
Thionin (Acetate)

Precaución: for Research Use Only

Formula: C₁₄H₁₃N₃O₂S
MDL Number: MFCD00081194
CAS Number: 78338-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR