Bis(dimethylamino)phosphoryl chloride 93%

Supplier: Ambeed
Danger

Synonyms: Phosphoric Acid Bis(dimethylamido) Chloride, N,N,N',N'-Tetramethylphosphorodiamidic Chloride, Bis(dimethylamino)phosphorochloridate, Bis(dimethylamido)phosphoric chloride

A286145-5G A286145-25G
76693-130EA 12 USD
76693-130 77601-258
Bis(dimethylamino)phosphoryl chloride 93%
Bis(dimethylamino)phosphoryl chloride

Precaución: for Research Use Only

Formula: C₄H₁₂ClN₂OP
MW: 170.58 g/mol
MDL Number: MFCD00008302
CAS Number: 1605-65-8
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR