Ammonium hexachlororuthenate(IV) 98%

Supplier: Ambeed
Danger

Synonyms: Diammonium hexachlororuthenate

A285251-250MG A285251-100MG A285251-1G
76693-126EA 19.1 USD
76693-126 77626-464 77626-466
Ammonium hexachlororuthenate(IV) 98%
Ammonium hexachlororuthenate(IV)

Precaución: for Research Use Only

Formula: H₈Cl₆N₂Ru
MW: 349.86 g/mol
MDL Number: MFCD00015962
CAS Number: 18746-63-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR