Benzofuran-5-sulfonyl chloride 95%

Supplier: Ambeed
Danger

A283214-100MG A283214-1G A283214-250MG A283214-5G
76693-096EA 90 USD
76693-096 76693-098 76693-100 76693-102
Benzofuran-5-sulfonyl chloride 95%
Benzofuran-5-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₈H₅ClO₃S
MDL Number: MFCD11053679
CAS Number: 869885-60-9
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR