Benzyltriethylammonium bromide 98%

Supplier: Ambeed
Warning

A282891-100G A282891-25G A282891-500G A282891-5G
76693-078EA 45 USD
76693-078 76693-080 76693-082 76693-084
Benzyltriethylammonium bromide 98%
Benzyltriethylammonium bromide

Precaución: for Research Use Only

Formula: C₁₃H₂₂BrN
MDL Number: MFCD00011822
CAS Number: 5197-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR