3-(3,4-Methylenedioxyphenyl)propionic acid 97%

Supplier: Ambeed
Warning

A282766-1G A282766-25G A282766-5G
76693-064EA 15 USD
76693-064 76693-066 76693-068
3-(3,4-Methylenedioxyphenyl)propionic acid 97%
3-(3,4-Methylenedioxyphenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MDL Number: MFCD00016909
CAS Number: 2815-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR