9-Phenyl-9-fluorenol 98%

Supplier: Ambeed
Warning

Synonyms: 9-Hydroxy-9-phenylfluorene, 9-Phenyl-9H-fluoren-9-ol

A280706-10G A280706-1G A280706-250MG A280706-5G
76693-016EA 72 USD
76693-016 76693-018 76693-020 76693-022
9-Phenyl-9-fluorenol 98%
9-Phenyl-9-fluorenol

Precaución: for Research Use Only

Formula: C₁₉H₁₄O
MDL Number: MFCD00037121
CAS Number: 25603-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR