2-(Carboxymethylthio)succinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-((Carboxymethyl)thio)succinic acid

A280329-1G A280329-5G
76693-010EA 25 USD
76693-010 76693-012
2-(Carboxymethylthio)succinic acid 97%
2-(Carboxymethylthio)succinic acid

Precaución: for Research Use Only

Formula: C₆H₈O₆S
MW: 208.184 g/mol
MDL Number: MFCD00002720
CAS Number: 99-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR