Cyclohexyl butyrate 98%

Supplier: Ambeed
Danger

A279207-25G A279207-5G
76692-988EA 9.55 USD
76692-988 76692-990
Cyclohexyl butyrate 98%
Cyclohexyl butyrate

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₂
MW: 170.25 g/mol
MDL Number: MFCD00046354
CAS Number: 1551-44-6
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR