2'-Iodoacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Iodophenyl)ethan-1-one, 1-(2-Iodophenyl)ethanone, o-Iodoacetophenone

A277189-100G A277189-10G A277189-1G A277189-25G A277189-500G A277189-5G
76692-958EA 119 USD
76692-958 76692-960 76692-962 76692-964 76692-966 76692-968
2'-Iodoacetophenone 98%
2'-Iodoacetophenone

Precaución: for Research Use Only

Formula: C₈H₇IO
MDL Number: MFCD00094998
CAS Number: 2142-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR