Methyl ((5S,10S,11S,14S)-11-benzyl-5-(tert-butyl)-10-hydroxy-15,15-dimethyl-3,6,13-trioxo-8-(4-(pyridin-2-yl)benzyl)-2-oxa-4,7,8,12-tetraazahexadecan-14-yl)carbamate sulfate 98%

Supplier: Ambeed
Warning

Synonyms: Atazanavir sulphate

A276573-10MG A276573-1G A276573-50MG A276573-25G
76692-952EA 5.31 USD
76692-952 76692-954 76692-956 77601-222
Methyl ((5S,10S,11S,14S)-11-benzyl-5-(tert-butyl)-10-hydroxy-15,15-dimethyl-3,6,13-trioxo-8-(4-(pyridin-2-yl)benzyl)-2-oxa-4,7,8,12-tetraazahexadecan-14-yl)carbamate sulfate 98%
Methyl ((5S,10S,11S,14S)-11-benzyl-5-(tert-butyl)-10-hydroxy-15,15-dimethyl-3,6,13-trioxo-8-(4-(pyridin-2-yl)benzyl)-2-oxa-4,7,8,12-tetraazahexadecan-14-yl)carbamate sulphate

Precaución: for Research Use Only

Formula: C₃₈H₅₄N₆O₁₁S
MW: 802.93 g/mol
MDL Number: MFCD08067748
CAS Number: 229975-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR