3-(4-Pyridyl)acrylic acid 98%

Supplier: Ambeed
Warning

A275095-100G A275095-10G A275095-1G A275095-25G A275095-5G
76692-936EA 201.58 USD
76692-936 76692-938 76692-940 76692-942 76692-944
3-(4-Pyridyl)acrylic acid 98%
3-(4-Pyridyl)acrylic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₂
MW: 149.15 g/mol
MDL Number: MFCD00040746
CAS Number: 5337-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR