4-Fluorophenetole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Fluorophenyl ethyl ether, 1-Fluoro-4-ethoxybenzene, 1-Ethoxy-4-fluorobenzene

A274891-1G A274891-25G A274891-5G
76692-930EA 8.49 USD
76692-930 76692-932 76692-934
4-Fluorophenetole 98%
4-Fluorophenetole

Precaución: for Research Use Only

Formula: C₈H₉FO
MW: 140.16 g/mol
MDL Number: MFCD00017953
CAS Number: 459-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR