(R)-(-)-4-Isopropyl-3-propionyl-2-oxazolidinone 98%

Supplier: Ambeed
Warning

A274307-1G A274307-250MG A274307-5G
76692-916EA 33 USD
76692-916 76692-918 76692-920
(R)-(-)-4-Isopropyl-3-propionyl-2-oxazolidinone 98%
(R)-(-)-4-Isopropyl-3-propionyl-2-oxazolidinone

Precaución: for Research Use Only

Formula: C₉H₁₅NO₃
MDL Number: MFCD00269691
CAS Number: 89028-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR