Ammonium hexachloroplatinate(IV) 98%

Supplier: Ambeed
Danger

Synonyms: Diammonium hexachloroplatinate

A274305-100MG A274305-1G A274305-250MG A274305-5G
76692-908EA 8 USD
76692-908 76692-910 76692-912 76692-914
Ammonium hexachloroplatinate(IV) 98%
Ammonium hexachloroplatinate(IV)

Precaución: for Research Use Only

Formula: H₈Cl₆N₂Pt
MW: 443.87
Density: 3.06
MDL Number: MFCD00010886
CAS Number: 16919-58-7
UN: 3288
ADR: 6.1 Ⅲ
Merck Index: 12,00582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR