7-Methoxy-1H-indole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A273673-100MG A273673-1G
76692-894EA 119.89 USD
76692-894 76692-896
7-Methoxy-1H-indole-2-carboxylic acid 95%
7-Methoxy-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃
MDL Number: MFCD06800927
CAS Number: 24610-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR