Primaquine diphosphate 98%

Supplier: Ambeed
Danger

Synonyms: N4-(6-Methoxyquinolin-8-yl)pentane-1,4-diamine bis(phosphate), NSC 149765, Primaquine bisphosphate

A273278-5G A273278-25G
76692-882EA 6.37 USD
76692-882 76692-884 77877-913
Primaquine diphosphate 98%
Primaquine diphosphate

Precaución: for Research Use Only

Formula: C₁₅H₂₇N₃O₉P₂
MW: 455.33 g/mol
MDL Number: MFCD00013166
CAS Number: 63-45-6
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR