Dihydromyricetin 98%

Supplier: Ambeed
Warning

Synonyms: (2R,3R)-3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-2,3-dihydrochromen-4-one, Ampelopsin, (2R,3R)-3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-4-one

A273270-10MG A273270-1G A273270-25G A273270-50MG A273270-5G A273270-5MG A273270-100G A273270-10G
76692-870EA 6 USD
76692-870 76692-872 76692-874 76692-876 76692-878 76692-880 77601-186 77601-188
Dihydromyricetin 98%
Dihydromyricetin

Precaución: for Research Use Only

Formula: C₁₅H₁₂O₈
MDL Number: MFCD00189451
CAS Number: 27200-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR