N-Acetyl-L-arginine dihydrate 97%

Supplier: Ambeed
Warning

A272745-100G A272745-25G A272745-5G
76692-858EA 181 USD
76692-858 76692-860 76692-862
N-Acetyl-L-arginine dihydrate 97%
N-Acetyl-L-arginine dihydrate

Precaución: for Research Use Only

Formula: C₈H₁₆N₄O₃·₂H₂O
MDL Number: MFCD00150285
CAS Number: 210545-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR