Cesium chloride 99%

Supplier: Ambeed
Warning

Synonyms: Cesium Chloride

A270389-100G A270389-25G A270389-500G
76692-834EA 182 USD
76692-834 76692-836 76692-838
Cesium chloride 99%
Cesium chloride

Precaución: for Research Use Only

Formula: CsCl
MW: 168.36 g/mol
Boiling Pt: 1303
Melting Pt: 646
MDL Number: MFCD00010955
CAS Number: 7647-17-8
Merck Index: 13,02021

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR