4-Fluoro-1H-indazol-7-amine 98%

Supplier: Ambeed
Warning

A270194-100MG A270194-1G A270194-250MG
76692-828EA 84.88 USD
76692-828 76692-830 76692-832
4-Fluoro-1H-indazol-7-amine 98%
4-Fluoro-1H-indazol-7-amine

Precaución: for Research Use Only

Formula: C₇H₆FN₃
MW: 151.14 g/mol
MDL Number: MFCD03787905
CAS Number: 866144-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR