5-Chlorobenzofuran-2-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Chlorobenzo[b]furan-2-carboxylic acid
A270039-1G
A270039-5G
76692-824EA
28.65
USD
76692-824
76692-826
5-Chlorobenzofuran-2-carboxylic acid 97%
5-Chlorobenzofuran-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₉H₅ClO₃ MW: 196.59 g/mol |
MDL Number:
MFCD00060512 CAS Number: 10242-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...