1,2-Bis(4-methoxyphenyl)ethyne 98%

Supplier: Ambeed
Warning

A269979-10G A269979-1G A269979-250MG A269979-5G
76692-816EA 1766.4 USD
76692-816 76692-818 76692-820 76692-822
1,2-Bis(4-methoxyphenyl)ethyne 98%
1,2-Bis(4-methoxyphenyl)ethyne

Precaución: for Research Use Only

Formula: C₁₆H₁₄O₂
MW: 238.28 g/mol
MDL Number: MFCD00025844
CAS Number: 2132-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR