Chromotropic acid disodium salt dihydrate 98,5%

Supplier: Ambeed
Warning

Synonyms: 4,5-Dihydroxynaphthalene-2,7-disulphonic acid disodium salt dihydrate, Disodium 4,5-dihydroxynaphthalene-2,7-disulphonate dihydrate, 4,5-Dihydroxynaphthalene-2,7-disulphonic acid, disodium salt dihydrate, 4,5-Dihydroxy-2,7-naphthalenedisulphonic acid disodium salt dihydrate

A269079-100G A269079-25G A269079-5G
76692-810EA 95.49 USD
76692-810 76692-812 76692-814
Chromotropic acid disodium salt dihydrate 98,5%
Chromotropic acid disodium salt dihydrate

Precaución: for Research Use Only

Formula: (HO)₂C₁₀H₄(SO₃Na)₂·2H₂O
MW: 400.29 g/mol
MDL Number: MFCD00150612
CAS Number: 5808-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR