8-Methylimidazo[1,2-a]pyridine 95%

Supplier: Ambeed
Warning

A267676-100MG A267676-1G A267676-250MG A267676-5G
76692-800EA 8.49 USD
76692-800 76692-802 76692-804 76692-806
8-Methylimidazo[1,2-a]pyridine 95%
8-Methylimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.1600 g/mol
MDL Number: MFCD05663805
CAS Number: 874-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR