2,4-Dichloro-3-ethyl-6-nitrophenol 98%

Supplier: Ambeed
Danger

A267601-25G A267601-5G
76692-796EA 20 USD
76692-796 76692-798
2,4-Dichloro-3-ethyl-6-nitrophenol 98%
2,4-Dichloro-3-ethyl-6-nitrophenol

Precaución: for Research Use Only

Formula: C₈H₇Cl₂NO₃
MW: 236.05 g/mol
MDL Number: MFCD00270764
CAS Number: 99817-36-4
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR