Ethyl-2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate

A267172-1G A267172-250MG A267172-5G
76692-786EA 30.77 USD
76692-786 76692-788 76692-790
Ethyl-2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate 95%
Ethyl-2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₈H₈F₃N₃O₂
MW: 235.17 g/mol
MDL Number: MFCD00052070
CAS Number: 149771-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR