1-(8-(Benzyloxy)-2-hydroxyquinolin-5-yl)-2-bromoethanone 97% (by HPLC)
Supplier: Ambeed
|
Danger
|
A266807-1G
A266807-25G
A266807-5G
76692-772EA
24.41
USD
76692-772
76692-774
76692-776
1-(8-(Benzyloxy)-2-hydroxyquinolin-5-yl)-2-bromoethanone 97% (by HPLC)
1-(8-(Benzyloxy)-2-hydroxyquinolin-5-yl)-2-bromoethanone
Precaución: for Research Use Only
Formula:
C₁₈H₁₄BrNO₃ MW: 372.21 g/mol |
MDL Number:
MFCD09025318 CAS Number: 100331-89-3 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...