Benzyltrimethylammonium bromide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N,N,N-Trimethyl-1-phenylmethanaminium bromide
A264654-100G
A264654-25G
A264654-500G
76692-752EA
45
USD
76692-752
76692-754
77877-883
Benzyltrimethylammonium bromide 97%
Benzyltrimethylammonium bromide
Precaución: for Research Use Only
Formula:
C₁₀H₁₆BrN |
MDL Number:
MFCD00011780 CAS Number: 5350-41-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...