Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) 98%

Supplier: Ambeed
Warning

Synonyms: Chlorocyclopentadienylbis(triphenylphosphine)ruthenium(II), Cyclopentadienylbis(triphenylphosphine)ruthenium(II) chloride, RuClCp(PPh3)

A264160-10G A264160-1G A264160-250MG A264160-5G A264160-25G
76692-740EA 66 USD
76692-740 76692-742 76692-744 76692-746 77626-388
Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) 98%
Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II)

Precaución: for Research Use Only

Formula: C₄₁H₃₅ClP₂Ru
MW: 726.20 g/mol
MDL Number: MFCD00075004
CAS Number: 32993-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR