Benzyl 2-acetamido-2-deoxy-α-D-glucopyranoside 95%

Supplier: Ambeed
Warning

Synonyms: Benzyl 2-(acetylamino)-2-deoxy-α-D-glucopyranoside, N-((2S,3R,4R,5S,6R)-2-(Benzyloxy)-4,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)acetamide

A263593-5G A263593-1G
76692-738EA 101.85 USD
76692-738 77571-630
Benzyl 2-acetamido-2-deoxy-α-D-glucopyranoside 95%
Benzyl 2-acetamido-2-deoxy-α-D-glucopyranoside

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₆
MW: 311.33 g/mol
MDL Number: MFCD00070371
CAS Number: 13343-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR