(S)-Benzyl 4-amino-2-((tert-butoxycarbonyl)amino)-4-oxobutanoate 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Asn-OBzl

A263486-10G A263486-1G A263486-5G
76692-732EA 240.83 USD
76692-732 76692-734 76692-736
(S)-Benzyl 4-amino-2-((tert-butoxycarbonyl)amino)-4-oxobutanoate 97%
(S)-Benzyl 4-amino-2-((tert-butoxycarbonyl)amino)-4-oxobutanoate

Precaución: for Research Use Only

Formula: C₁₆H₂₂N₂O₅
MW: 322.35 g/mol
MDL Number: MFCD00190784
CAS Number: 13512-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR