Clopidogrel bisulfate 98%

Supplier: Ambeed
Warning

Synonyms: (±)-Clopidogrel hydrogen sulphate, Methyl 2-(2-chlorophenyl)-2-(6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl)acetate sulphate, (±)-Clopidogrel bisulphate

A262835-1G A262835-5G
76692-724EA 11 USD
76692-724 77571-620
Clopidogrel bisulfate 98%
Clopidogrel bisulfate

Precaución: for Research Use Only

Formula: C₁₆H₁₈ClNO₆S₂
MDL Number: MFCD05865229
CAS Number: 135046-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR