2-(Benzyloxy)acetaldehyde ≥95% stabilized

Supplier: Ambeed
Warning

Synonyms: (Benzyloxy)acetaldehyde, Benzyloxyacetaldehyde

Stabilization: Stabilized with Catechol 0.5%

A261490-10G A261490-25G A261490-5G A261490-100G
76692-710EA 98.67 USD
76692-710 76692-712 76692-714 77568-510
2-(Benzyloxy)acetaldehyde ≥95% stabilized
2-(Benzyloxy)acetaldehyde

Precaución: For research use only.

Formula: C₉H₁₀O₂
MW: 150.177 g/mol
MDL Number: MFCD00191779
CAS Number: 60656-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR