Methyl-4-hydroxy-3,5-dimethoxybenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-Hydroxy-3,5-Dimethoxybenzoate, Methyl syringate

A260750-100G A260750-100MG A260750-10G A260750-1G A260750-25G A260750-5G
76692-690EA 133 USD
76692-690 76692-692 76692-694 76692-696 76692-698 76692-700
Methyl-4-hydroxy-3,5-dimethoxybenzoate 97%
Methyl-4-hydroxy-3,5-dimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₅
MDL Number: MFCD00017199
CAS Number: 884-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR