Amyl-2-methylbutyrate 98%

Supplier: Ambeed
Warning

A260348-25G A260348-5G
76692-688EA 28.65 USD
76692-688 77626-356
Amyl-2-methylbutyrate 98%
Amyl-2-methylbutyrate

Precaución: for Research Use Only

Formula: C₁₀H₂₀O₂
MW: 172.26 g/mol
MDL Number: MFCD00129713
CAS Number: 68039-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR