Cycloheptanol 98%

Supplier: Ambeed
Warning

Synonyms: Hydroxycycloheptane, Suberyl alcohol

A259418-100G A259418-1G A259418-5G A259418-25G
76692-674EA 176.11 USD
76692-674 76692-676 76692-678 77626-350
Cycloheptanol 98%
Cycloheptanol

Precaución: for Research Use Only

Formula: C₇H₁₄O
MW: 114.19 g/mol
MDL Number: MFCD00004150
CAS Number: 502-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR