Inosine 98%

Supplier: Ambeed
Warning

Synonyms: 9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one

A257563-100G A257563-25G A257563-500G A257563-1000G
76692-642EA 9.55 USD
76692-642 76692-644 76692-646 77593-612
Inosine 98%
Inosine

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₄O₅
MW: 268.23 g/mol
MDL Number: MFCD00066770
CAS Number: 58-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR