8-(4-Bromophenyl)-1,4-dioxaspiro[4.5]decan-8-ol 98+%

Supplier: Ambeed
Warning

A254715-100MG A254715-1G A254715-250MG
76692-592EA 38.2 USD
76692-592 76692-594 76692-596
8-(4-Bromophenyl)-1,4-dioxaspiro[4.5]decan-8-ol 98+%
8-(4-Bromophenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: for Research Use Only

Formula: C₁₄H₁₇BrO₃
MW: 313.18 g/mol
MDL Number: MFCD06410871
CAS Number: 125962-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR