Benzyltrimethylammonium tetrachloroiodate 98%

Supplier: Ambeed
Danger

A254218-1G A254218-250MG A254218-5G
76692-580EA 44.56 USD
76692-580 76692-582 76692-584
Benzyltrimethylammonium tetrachloroiodate 98%
Benzyltrimethylammonium tetrachloroiodate

Precaución: for Research Use Only

Formula: C₁₀H₁₆Cl₄IN
MW: 418.95 g/mol
MDL Number: MFCD00191607
CAS Number: 121309-88-4
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR