Benzyl butyrate 98%

Supplier: Ambeed
Warning

Synonyms: Butyric acid benzyl ester, Butanoic acid benzyl ester

A253856-1000G A253856-100G A253856-500G
76692-568EA 42.44 USD
76692-568 76692-570 76692-572
Benzyl butyrate 98%
Benzyl butyrate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
MDL Number: MFCD00027133
CAS Number: 103-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR