cis-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: cis-3-(Boc-amino)cyclohexanecarboxylic Acid, cis-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid, cis-3-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid

A253750-1G A253750-5G A253750-25G A253750-10G
76692-564EA 12.74 USD
76692-564 76692-566 77601-038 77626-330
cis-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid 97%
cis-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MW: 243.30 g/mol
MDL Number: MFCD03453252
CAS Number: 222530-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR