2,5-Dimethylterephthalic acid 98%

Supplier: Ambeed
Warning

A253470-100MG A253470-1G A253470-250MG A253470-5G
76692-556EA 11 USD
76692-556 76692-558 76692-560 76692-562
2,5-Dimethylterephthalic acid 98%
2,5-Dimethylterephthalic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MDL Number: MFCD00020292
CAS Number: 6051-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR