Ethyl-2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-5,6-dihydro-4-[4H]-cyclopenta[B]thiophene-3-carboxylic acid ethyl ester, Ethyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
A252546-100G
A252546-10G
A252546-1G
A252546-250MG
A252546-25G
A252546-5G
76692-528EA
381.93
USD
76692-528
76692-530
76692-532
76692-534
76692-536
76692-538
Ethyl-2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate 96%
Ethyl-2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₃NO₂S MW: 211.28 g/mol |
MDL Number:
MFCD00102063 CAS Number: 4815-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...