Methyl-2-amino-5-bromobenzoate 98%
Supplier: Ambeed
|
Warning
|
A251987-100G
A251987-10G
A251987-1G
A251987-25G
A251987-5G
A251987-500G
76692-492EA
59
USD
76692-492
76692-494
76692-496
76692-498
76692-500
77550-598
Methyl-2-amino-5-bromobenzoate 98%
Methyl-2-amino-5-bromobenzoate
Precaución: for Research Use Only
Formula:
C₈H₈BrNO₂ |
MDL Number:
MFCD00144761 CAS Number: 52727-57-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...