2,3,5,6-Tetramethyl-m-phenylenediamine 98%

Supplier: Ambeed
Warning

Synonyms: 3,6-Diaminodurene, 2,3,5,6-Tetramethylbenzene-1,3-diamine, 3,6-Diamino-1,2,4,5-tetramethylbenzene, 1,4-Diamino-2,3,5,6-tetramethylbenzene, 2,3,5,6-Tetramethyl-1,4-phenylenediamine, 2,3,5,6-Tetramethyl-1,3-phenylenediamine

A251537-1G A251537-250MG A251537-25G A251537-5G
76692-478EA 14 USD
76692-478 76692-480 76692-482 76692-484
2,3,5,6-Tetramethyl-m-phenylenediamine 98%
2,3,5,6-Tetramethyl-m-phenylenediamine

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂
MDL Number: MFCD00007905
CAS Number: 3102-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR