Quinoline-N-oxide 98%

Supplier: Ambeed
Warning

Synonyms: Quinoline 1-oxide, Quinoline-1-oxide

A250595-100G A250595-1G A250595-5G
76692-472EA 422.24 USD
76692-472 76692-474 76692-476
Quinoline-N-oxide 98%
Quinoline-N-oxide

Precaución: for Research Use Only

Formula: C₉H₇NO
MW: 145.161 g/mol
MDL Number: MFCD00006737
CAS Number: 1613-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR