1-Cetylpyridinium chloride 97%

Supplier: Ambeed
Warning

Synonyms: Cetylpyridinium chloride, 1-Hexadecylpyridinium chloride, 1-Hexadecylpyridin-1-ium chloride

A249533-100G A249533-1G A249533-5G A249533-25G A249533-500G
76692-434EA 50 USD
76692-434 76692-436 76692-438 77601-012 77626-312
1-Cetylpyridinium chloride 97%
1-Cetylpyridinium chloride

Precaución: for Research Use Only

Formula: C₂₁H₃₈ClN
MDL Number: MFCD00011731
CAS Number: 123-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR