β-Cyclodextrine 97%

Supplier: Ambeed
Warning

Synonyms: Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex

A248973-1000G A248973-100G A248973-25G A248973-500G
76692-422EA 39.26 USD
76692-422 76692-424 76692-426 76692-428
β-Cyclodextrine 97%
β-Cyclodextrine

Precaución: for Research Use Only

Formula: C₄₂H₇₀O₃₅
MW: 1134.98 g/mol
MDL Number: MFCD00078139
CAS Number: 7585-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR